Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Biphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
Packaging:
AcroSeal™ Glass Bottle
3315-91-1,109-99-9
C12H9BrMg
257.41
JWQLJPBJNSPKSG-UHFFFAOYSA-M
4-biphenylmagnesium bromide solution,magnesium 2+ ion 4-phenylbenzen-1-ide bromide
54721138
magnesium;phenylbenzene;bromide
C1=CC=C(C=C1)C2=CC=[C-]C=C2.[Mg+2].[Br-]
3315-91-1,109-99-9
C12H9BrMg
257.41
JWQLJPBJNSPKSG-UHFFFAOYSA-M
4-biphenylmagnesium bromide solution,magnesium 2+ ion 4-phenylbenzen-1-ide bromide
54721138
magnesium;phenylbenzene;bromide
C1=CC=C(C=C1)C2=CC=[C-]C=C2.[Mg+2].[Br-]
Specific Gravity 0.961
Packaging AcroSeal™ Glass Bottle
Flash Point −17°C
Quantity 100mL
Color Yellow to Brown
Density 0.9610g/mL
Boiling Point 65.0°C
CAS Min % 10.71,83.93
CAS Max % 16.07,89.29
Formula Weight 257.41
Physical Form Liquid
Chemical Name or Material 4-Biphenylmagnesium bromide
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger