Alfa Aesar™ 4-[Bis(2-methoxyethyl)aminomethyl]benzeneboronic acid pinacol ester, 95%

Manufacturer: Alfa Aesar™ H5194203

Quantity 1g

 View more versions of this product

Catalog No. AAH5194203

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C19H32BNO4
    Formula Weight 349.28
    MDL Number MFCD16294545
    Synonym Bis(2-methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzylamine
    Chemical Name or Material 4-[Bis(2-methoxyethyl)aminomethyl]benzeneboronic acid pinacol ester

    Specifications

    Formula Weight 349.28
    Molecular Formula C19H32BNO4
    MDL Number MFCD16294545
    Synonym Bis(2-methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzylamine
    Quantity 1g
    Chemical Name or Material 4-[Bis(2-methoxyethyl)aminomethyl]benzeneboronic acid pinacol ester
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit