Alfa Aesar™ 4-Boc-1-(5-bromo-2-pyridyl)piperazine, 97%

Manufacturer: Alfa Aesar™ H5531114

Quantity 25g

 View more versions of this product

Catalog No. AAH5531114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 153747-97-8
    Formula Weight 342.23
    MDL Number MFCD07369772
    UN Number UN2811
    Synonym 2-(1-Boc-1-piperazinyl)-5-bromopyridine
    Chemical Name or Material 4-Boc-1-(5-bromo-2-pyridyl)piperazine

    Specifications

    Melting Point 83° to 87°C
    Formula Weight 342.23
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Color White to yellow
    Synonym 2-(1-Boc-1-piperazinyl)-5-bromopyridine
    MDL Number MFCD07369772
    CAS 153747-97-8
    UN Number UN2811
    Physical Form Powder
    Quantity 25g
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material 4-Boc-1-(5-bromo-2-pyridyl)piperazine
    Assay 97%

    Description

    Applications
    4-Boc-1-(5-bromo-2-pyridyl)piperazine is used as active pharmaceutical ingredient.

    Solubility
    Soluble in water.

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit