Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Boc-amino)-N-Fmoc-D-phenylalanine, 95%

Click to view available options
Quantity:
250mg
1g
5g
214750-77-3
C29H30N2O6
502.567
MFCD00798642
KVUAOWDVYMUKPE-RUZDIDTESA-N
fmoc-d-phe 4-nhboc-oh,fmoc-4-boc-amino-d-phenylalanine,fmoc-d-4-aminophe boc,fmoc-d-4-aminophenylalanine boc,2r-3-4-tert-butoxycarbonyl amino phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid,fmoc-p-amino-phe boc-oh,fmoc-d-phe boc-4-nh2-oh,fmoc-p-amino-d-phe boc-oh,4-boc-amino-n-fmoc-d-phenylalanine,2r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-4-2-methylpropan-2-yl oxycarbonylamino phenyl propanoic acid
7021127
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]propanoic acid
CC(C)(C)OC(=O)NC1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
214750-77-3
C29H30N2O6
502.567
MFCD00798642
KVUAOWDVYMUKPE-RUZDIDTESA-N
fmoc-d-phe 4-nhboc-oh,fmoc-4-boc-amino-d-phenylalanine,fmoc-d-4-aminophe boc,fmoc-d-4-aminophenylalanine boc,2r-3-4-tert-butoxycarbonyl amino phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid,fmoc-p-amino-phe boc-oh,fmoc-d-phe boc-4-nh2-oh,fmoc-p-amino-d-phe boc-oh,4-boc-amino-n-fmoc-d-phenylalanine,2r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-4-2-methylpropan-2-yl oxycarbonylamino phenyl propanoic acid
7021127
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]propanoic acid
CC(C)(C)OC(=O)NC1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Formula Weight 502.57
Physical Form Solid
Percent Purity 95%
Quantity 250mg
Chemical Name or Material 4-(Boc-amino)-N-Fmoc-D-phenylalanine

storageNote1 : -30° to -10°C