Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Boc-aminomethyl)benzoic acid, 98%

Click to view available options
Quantity:
5g
25g
33233-67-9
C13H17NO4
251.282
MFCD00228182
LNKHBRDWRIIROP-UHFFFAOYSA-N
4-tert-butoxycarbonylamino methyl benzoic acid,4-tert-butoxycarbonyl amino methyl benzoic acid,4-tert-butoxycarbonyl amino methyl benzoic acid,4-tert-butoxy carbonyl amino methyl benzoic acid,boc-4-aminomethyl-benzoic acid,4-boc-aminomethyl-benzoic acid,4-tert-butoxycarbonylaminomethyl benzoic acid,4-n-tert-butyloxycarbonyl amino methyl benzoic acid,benzoic acid, 4-1,1-dimethylethoxy carbonyl amino methyl,4-tert-butoxycarbonylamino methyl benzoic acid
853605
4-[[(2-methylpropan-2-yl)oxycarbonylamino]methyl]benzoic acid
CC(C)(C)OC(=O)NCC1=CC=C(C=C1)C(=O)O
33233-67-9
C13H17NO4
251.282
MFCD00228182
LNKHBRDWRIIROP-UHFFFAOYSA-N
4-tert-butoxycarbonylamino methyl benzoic acid,4-tert-butoxycarbonyl amino methyl benzoic acid,4-tert-butoxycarbonyl amino methyl benzoic acid,4-tert-butoxy carbonyl amino methyl benzoic acid,boc-4-aminomethyl-benzoic acid,4-boc-aminomethyl-benzoic acid,4-tert-butoxycarbonylaminomethyl benzoic acid,4-n-tert-butyloxycarbonyl amino methyl benzoic acid,benzoic acid, 4-1,1-dimethylethoxy carbonyl amino methyl,4-tert-butoxycarbonylamino methyl benzoic acid
853605
4-[[(2-methylpropan-2-yl)oxycarbonylamino]methyl]benzoic acid
CC(C)(C)OC(=O)NCC1=CC=C(C=C1)C(=O)O
Color White
Quantity 5g
Melting Point 164°C to 168°C
Beilstein 2854286
Formula Weight 251.28
Physical Form Crystalline
Percent Purity 98%
Chemical Name or Material 4-(Boc-aminomethyl)benzoic acid