Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Borono-D-phenylalanine, 97%, Acros Organics

Click to view available options
Quantity:
100mg
Packaging:
Glass bottle
111821-49-9
C9H12BNO4
209.008
NFIVJOSXJDORSP-MRVPVSSYSA-N
4-borono-d-phenylalanine,r-2-amino-3-4-boronophenyl propanoic acid,d-phenylalanine, 4-borono,2r-2-amino-3-4-boronophenyl propanoic acid,2r-2-amino-3-4-dihydroxyboranyl phenyl propanoic acid,phenylalanine, 4-borono,4-dihydroxyboryl phenylalanine,pubchem7900,4-dihydroxyboryl-d-phenylalanine
2734317
(2R)-2-amino-3-(4-boronophenyl)propanoic acid
B(C1=CC=C(C=C1)CC(C(=O)O)N)(O)O
111821-49-9
C9H12BNO4
209.008
NFIVJOSXJDORSP-MRVPVSSYSA-N
4-borono-d-phenylalanine,r-2-amino-3-4-boronophenyl propanoic acid,d-phenylalanine, 4-borono,2r-2-amino-3-4-boronophenyl propanoic acid,2r-2-amino-3-4-dihydroxyboranyl phenyl propanoic acid,phenylalanine, 4-borono,4-dihydroxyboryl phenylalanine,pubchem7900,4-dihydroxyboryl-d-phenylalanine
2734317
(2R)-2-amino-3-(4-boronophenyl)propanoic acid
B(C1=CC=C(C=C1)CC(C(=O)O)N)(O)O
Quantity 100mg
Packaging Glass bottle
Assay Percent Range 97%
Formula Weight 209.01
Chemical Name or Material 4-Borono-D-phenylalanine