Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 4-Bromo-1,1,2-trifluoro-1-butene, 98%
Click to view available options
Quantity:
10g
50g
Description
Applications
4-Bromo-1,1,2-trifluoro-1-butene is used in organic synthesis, chemical research and pharmaceuticals.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
4-Bromo-1,1,2-trifluoro-1-butene is used in organic synthesis, chemical research and pharmaceuticals.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
Specifications
Sensitivity | Light sensitive |
Refractive Index | 1.402 |
Density | 1.639 |
Odor | Recognizable |
Boiling Point | 95°C to 98°C |
Quantity | 10g |
Flash Point | 17°C (62°F) |
Linear Formula | BrCH2CH2CF=CF2 |
UN Number | UN1993 |
Beilstein | 1840755 |
Show More |
Safety and Handling
GHS H Statement
H225-H319-H335
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 234-019-2
TSCA : No
Recommended Storage : Ambient temperatures