Alfa Aesar™ 4-Bromo-1,2-dichlorobenzene, 98+%

Manufacturer: Alfa Aesar™ A1031406

Quantity 5g

 View more versions of this product

Catalog No. AAA1031406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18282-59-2
    Molecular Formula C6H3BrCl2
    Formula Weight 225.91
    MDL Number MFCD00040849
    Beilstein 1635735
    Synonym 1-Bromo-3,4-dichlorobenzene; 3,4-Dichlorobromobenzene
    Chemical Name or Material 4-Bromo-1,2-dichlorobenzene

    Specifications

    Formula Weight 225.91
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Quantity 5g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Molecular Formula C6H3BrCl2
    MDL Number MFCD00040849
    CAS 18282-59-2
    Melting Point 22° to 25°C
    Synonym 1-Bromo-3,4-dichlorobenzene; 3,4-Dichlorobromobenzene
    Boiling Point 140° to 142°C (30mmHg)
    Beilstein 1635735
    Flash Point 87°C (188°F)
    Chemical Name or Material 4-Bromo-1,2-dichlorobenzene
    Assay ≥98%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit