Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-1-butanol, 85+%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
33036-62-3
C4H9BrO
153.02
MFCD00015387
SIJLYRDVTMMSIP-UHFFFAOYSA-N
4-bromo-1-butanol,1-butanol, 4-bromo,4-bromobutanol,4-bromo-butan-1-ol,1-bromo-4-hydroxybutane,tetramethylene bromohydrin,4-bromo-n-butanol,1-bromobutan-4-ol,4-bromo butan-1-ol,acmc-1axcv
118388
4-bromobutan-1-ol
C(CCBr)CO
33036-62-3
C4H9BrO
153.02
MFCD00015387
SIJLYRDVTMMSIP-UHFFFAOYSA-N
4-bromo-1-butanol,1-butanol, 4-bromo,4-bromobutanol,4-bromo-butan-1-ol,1-bromo-4-hydroxybutane,tetramethylene bromohydrin,4-bromo-n-butanol,1-bromobutan-4-ol,4-bromo butan-1-ol,acmc-1axcv
118388
4-bromobutan-1-ol
C(CCBr)CO
Density 1.0680g/mL
Color Colorless to Brown
Specific Gravity 1.068
Quantity 1g
Boiling Point 56.0°C to 58.0°C (2.0mmHg)
Flash Point 7°C
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 85.0
CAS Max % 100.0
Assay Percent Range 85% min. (GC)
Linear Formula Br(CH2)4OH
Solubility Information Solubility in water: insoluble
Formula Weight 153.02
Physical Form Liquid
Percent Purity 85+%
Chemical Name or Material 4-Bromo-1-butanol
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 251-355-5