Alfa Aesar™ 4-Bromo-1-butanol, tech., cont. varying amounts of THF

Manufacturer: Alfa Aesar™ 1680514

Quantity 25g

 View more versions of this product

Catalog No. AA1680514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 33036-62-3
    Linear Formula Br(CH2)4OH
    Formula Weight 153.02
    MDL Number MFCD00015387
    UN Number UN1987
    Chemical Name or Material 4-Bromo-1-butanol, cont. varying amounts of THF
    Grade Technical Grade

    Specifications

    Formula Weight 153.02
    Health Hazard 3 GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Sensitivity Moisture & Light Sensitive
    Health Hazard 1 Warning
    Storage Note 1 −30°C to −10°C
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
    Boiling Point 63° to 65°C (5mmHg)
    Physical Form Liquid
    MDL Number MFCD00015387
    UN Number UN1987
    Refractive Index 1.5035
    Color Yellow
    Quantity 25g
    Flash Point 32° (91°F)
    Linear Formula Br(CH2)4OH
    CAS 33036-62-3
    Health Hazard 2 GHS H Statement
    H226-H351-H315-H319
    Flammable liquid and vapor.
    Suspected of causing cancer.
    Causes skin irritation.
    Causes serious eye irritation.
    Chemical Name or Material 4-Bromo-1-butanol, cont. varying amounts of THF
    Grade Technical Grade

    GHS H Statement
    H226-H351-H315-H319
    Flammable liquid and vapor.
    Suspected of causing cancer.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    −30°C to −10°C

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit