Learn More
4-Bromo-1-butene, 97%, Thermo Scientific Chemicals
Description
4-Bromo-1-butene is used in the preparation of 4-(3-butenyloxy)benzoic acid and largazole. It is also used in double alkylation of lactams followed by Grubbs ring-closing metathesis to yield gymnodimine. Further, it serves as a reagent in the preparation of dienehydrazides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Bromo-1-butene is used in the preparation of 4-(3-butenyloxy)benzoic acid and largazole. It is also used in double alkylation of lactams followed by Grubbs ring-closing metathesis to yield gymnodimine. Further, it serves as a reagent in the preparation of dienehydrazides.
Solubility
Miscible with water, benzene, ether and ethanol.
Notes
Light sensitive. Store in a cool place. Incompatible with strong bases.
Specifications
Density | 1.33 |
Boiling Point | 98°C to 100°C |
Flash Point | 1°C (34°F) |
Refractive Index | 1.463 |
Quantity | 25 g |
UN Number | UN1993 |
Beilstein | 1098377 |
Sensitivity | Light sensitive |
Solubility Information | Miscible with water,benzene,ether and ethanol. |
SMILES | C=CCCBr |
Show More |
Safety and Handling
Hazard Category | H225-H317-H319-H334-H335 |
Hazard Statement | GHS H Statement H225-H334-H335-H319-H317 Highly flammable liquid and vapor. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause respiratory irritation. Causes serious eye irritation. May cause an allergic skin reaction. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P333+P313-P342+P311-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
EINECSNumber | 225-937-4 |
TSCA | Yes |
Recommended Storage | Keep cold |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.