Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-1-chloro-2-fluorobenzene, 98%

Click to view available options
Quantity:
5g
25g
100g
60811-18-9
C6H3BrClF
209.442
MFCD00042519
AGYWDGVTLKNTBS-UHFFFAOYSA-N
1-bromo-4-chloro-3-fluorobenzene,3-fluoro-4-chlorobromobenzene,2-fluoro-4-bromochlorobenzene,2-chloro-5-bromofluorobenzene,4-chloro-3-fluorobromobenzene,5-bromo-2-chlorofluorobenzene,4-bromo-1-chloro-2-fluoro-benzene,4-bromo-2-fluorochlorobenzene,benzene, 4-bromo-1-chloro-2-fluoro,3-fluoro-4-chloro-bromobenzene
2724690
4-bromo-1-chloro-2-fluorobenzene
C1=CC(=C(C=C1Br)F)Cl
60811-18-9
C6H3BrClF
209.442
MFCD00042519
AGYWDGVTLKNTBS-UHFFFAOYSA-N
1-bromo-4-chloro-3-fluorobenzene,3-fluoro-4-chlorobromobenzene,2-fluoro-4-bromochlorobenzene,2-chloro-5-bromofluorobenzene,4-chloro-3-fluorobromobenzene,5-bromo-2-chlorofluorobenzene,4-bromo-1-chloro-2-fluoro-benzene,4-bromo-2-fluorochlorobenzene,benzene, 4-bromo-1-chloro-2-fluoro,3-fluoro-4-chloro-bromobenzene
2724690
4-bromo-1-chloro-2-fluorobenzene
C1=CC(=C(C=C1Br)F)Cl
Boiling Point 62°C to 64°C (9mmHg)
Quantity 5g
Refractive Index 1.555
Density 1.71
Beilstein 2206649
Formula Weight 209.45
Percent Purity 98%
Chemical Name or Material 4-Bromo-1-chloro-2-fluorobenzene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures