Alfa Aesar™ 4-Bromo-1-fluoro-2-nitrobenzene, 98%

Manufacturer: Alfa Aesar™ B2252314

Quantity 25g

 View more versions of this product

Catalog No. AAB2252314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 364-73-8
    Molecular Formula C6H3BrFNO2
    Formula Weight 220.01
    MDL Number MFCD00129165
    UN Number UN2810
    Beilstein 2329084
    Synonym 5-Bromo-2-fluoronitrobenzene; 1-Bromo-4-fluoro-3-nitrobenzene
    Chemical Name or Material 4-Bromo-1-fluoro-2-nitrobenzene

    Specifications

    Formula Weight 220.01
    Molecular Formula C6H3BrFNO2
    MDL Number MFCD00129165
    UN Number UN2810
    Melting Point 18° to 19°C
    Synonym 5-Bromo-2-fluoronitrobenzene; 1-Bromo-4-fluoro-3-nitrobenzene
    Boiling Point 240° to 241°C
    Quantity 25g
    Flash Point >110°C (230°F)
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 364-73-8
    Beilstein 2329084
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Chemical Name or Material 4-Bromo-1-fluoro-2-nitrobenzene
    Assay 98%

    Description

    Applications
    4-Bromo-1-fluoro-2-nitrobenzene is used in the synthesis of anti-inflammatory agents.

    Solubility
    Difficult to mix.

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H331-H302-H312-H315-H319
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit