Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-1-methyl-1H-indole, 95%

Click to view available options
Quantity:
250mg
1g
590417-55-3
C9H8BrN
210.074
MFCD08435875
JZOSXTYDJPHXQD-UHFFFAOYSA-N
4-bromo-1-methyl-1h-indole,4-bromo-1-methyl-indole,acmc-209m9m,1h-indole,4-bromo-1-methyl,4-bromo-1-methylindole,1h-indole, 4-bromo-1-methyl,4-bromo-1-methyl-1h-indole
22558937
4-bromo-1-methylindole
CN1C=CC2=C1C=CC=C2Br
590417-55-3
C9H8BrN
210.074
MFCD08435875
JZOSXTYDJPHXQD-UHFFFAOYSA-N
4-bromo-1-methyl-1h-indole,4-bromo-1-methyl-indole,acmc-209m9m,1h-indole,4-bromo-1-methyl,4-bromo-1-methylindole,1h-indole, 4-bromo-1-methyl,4-bromo-1-methyl-1h-indole
22558937
4-bromo-1-methylindole
CN1C=CC2=C1C=CC=C2Br
Refractive Index 1.6360
Quantity 1g
Density 1.456g/mL
Color Colorless to Yellow
Formula Weight 210.06
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 4-Bromo-1-methyl-1H-indole