Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-1-methyl-1H-pyrazole-3-carboxaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
287917-96-8
C5H5BrN2O
189.01
KGIYMMPLYIITLL-UHFFFAOYSA-N
4-bromo-1-methyl-1h-pyrazole-3-carbaldehyde,4-bromo-3-formyl-1-methyl-1h-pyrazole,4-bromo-1-methylpyrazole-3-carboxaldehyde,4-bromo-1-methyl-1h-pyrazole-3-carboxaldehyde,1h-pyrazole-3-carboxaldehyde, 4-bromo-1-methyl,pubchem23668,acmc-209h4p,buttpark 88\08-06,4-bromo-3-formyl-1-methylpyrazole,1h-pyrazole-3-carboxaldehyde,4-bromo-1-methyl
2735595
4-bromo-1-methylpyrazole-3-carbaldehyde
CN1C=C(C(=N1)C=O)Br
287917-96-8
C5H5BrN2O
189.01
KGIYMMPLYIITLL-UHFFFAOYSA-N
4-bromo-1-methyl-1h-pyrazole-3-carbaldehyde,4-bromo-3-formyl-1-methyl-1h-pyrazole,4-bromo-1-methylpyrazole-3-carboxaldehyde,4-bromo-1-methyl-1h-pyrazole-3-carboxaldehyde,1h-pyrazole-3-carboxaldehyde, 4-bromo-1-methyl,pubchem23668,acmc-209h4p,buttpark 88\08-06,4-bromo-3-formyl-1-methylpyrazole,1h-pyrazole-3-carboxaldehyde,4-bromo-1-methyl
2735595
4-bromo-1-methylpyrazole-3-carbaldehyde
CN1C=C(C(=N1)C=O)Br
Melting Point 71°C to 76°C
Packaging Glass Bottle
Quantity 1g
Assay Percent Range 96% min. (GC)
Formula Weight 189.01
Percent Purity 97%
Chemical Name or Material 4-Bromo-1-methyl-1H-pyrazole-3-carboxaldehyde

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e