Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-1-methyl-1H-pyrazole, ≥98%

Click to view available options
Quantity:
25g
100g
15803-02-8
C4H5BrN2
161.002
MFCD02179565
IXJSDKIJPVSPKF-UHFFFAOYSA-N
4-bromo-1-methyl-1h-pyrazole,1-methyl-4-bromo-1h-pyrazole,1h-pyrazole, 4-bromo-1-methyl,1-mehtyl-4-bromo-1h-pyrazole,1-methyl-4-bromopyrazole,4-bromo-1-methyl-pyrazole,4-bromo-2-methyl-1h-pyrazole,zlchem 151,pubchem16867,4bmp
167433
4-bromo-1-methylpyrazole
CN1C=C(C=N1)Br
15803-02-8
C4H5BrN2
161.002
MFCD02179565
IXJSDKIJPVSPKF-UHFFFAOYSA-N
4-bromo-1-methyl-1h-pyrazole,1-methyl-4-bromo-1h-pyrazole,1h-pyrazole, 4-bromo-1-methyl,1-mehtyl-4-bromo-1h-pyrazole,1-methyl-4-bromopyrazole,4-bromo-1-methyl-pyrazole,4-bromo-2-methyl-1h-pyrazole,zlchem 151,pubchem16867,4bmp
167433
4-bromo-1-methylpyrazole
CN1C=C(C=N1)Br
Boiling Point 185°C to 188°C
Color Colorless to Yellow
Refractive Index 1.5310
Quantity 25g
Density 1.558g/mL
Beilstein 108120
Formula Weight 161
Physical Form Liquid
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-1-methyl-1H-pyrazole

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger