Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-1-naphthoic acid, 98%

Click to view available options
Quantity:
1g
5g
16650-55-8
C11H7BrO2
251.079
MFCD00094221
FIJIPZQZVLCOMB-UHFFFAOYSA-N
4-bromo-1-naphthoic acid,4-bromo-1-naphthalenecarboxylic acid,4-bromo-1-naphtalenecarboxylic acid,1-naphthalenecarboxylic acid, 4-bromo,4-bromo naphthoic acid,acmc-209dum,4-bromo-naphthalic acid,4-bromo-1-naphthoicacid,ksc175i3p,1-naphthalenecarboxylicacid,4-bromo
282591
4-bromonaphthalene-1-carboxylic acid
C1=CC=C2C(=C1)C(=CC=C2Br)C(=O)O
16650-55-8
C11H7BrO2
251.079
MFCD00094221
FIJIPZQZVLCOMB-UHFFFAOYSA-N
4-bromo-1-naphthoic acid,4-bromo-1-naphthalenecarboxylic acid,4-bromo-1-naphtalenecarboxylic acid,1-naphthalenecarboxylic acid, 4-bromo,4-bromo naphthoic acid,acmc-209dum,4-bromo-naphthalic acid,4-bromo-1-naphthoicacid,ksc175i3p,1-naphthalenecarboxylicacid,4-bromo
282591
4-bromonaphthalene-1-carboxylic acid
C1=CC=C2C(=C1)C(=CC=C2Br)C(=O)O
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Quantity 5g
Beilstein 1950351
Formula Weight 251.08
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 4-Bromo-1-naphthoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning