Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-1-trityl-1H-pyrazole, 95%

Click to view available options
Quantity:
1g
5g
95162-14-4
C22H17BrN2
389.296
MFCD09907863
CPENTLJGGGSVAJ-UHFFFAOYSA-N
4-bromo-1-trityl-1h-pyrazole,1h-pyrazole, 4-bromo-1-triphenylmethyl,1h-pyrazole,4-bromo-1-triphenylmethyl,4-bromo-1-triphenylmethyl-1h-pyrazole,4-bromo-1-triphenylmethyl pyrazole,n-trityl-4-bromopyrazole,4-bromo-1-n-tritylpyrazole,1-trityl-4-bromo-1h-pyrazole,4-bromo-1-trityl-1-h-pyrazole
10834188
4-bromo-1-tritylpyrazole
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=C(C=N4)Br
95162-14-4
C22H17BrN2
389.296
MFCD09907863
CPENTLJGGGSVAJ-UHFFFAOYSA-N
4-bromo-1-trityl-1h-pyrazole,1h-pyrazole, 4-bromo-1-triphenylmethyl,1h-pyrazole,4-bromo-1-triphenylmethyl,4-bromo-1-triphenylmethyl-1h-pyrazole,4-bromo-1-triphenylmethyl pyrazole,n-trityl-4-bromopyrazole,4-bromo-1-n-tritylpyrazole,1-trityl-4-bromo-1h-pyrazole,4-bromo-1-trityl-1-h-pyrazole
10834188
4-bromo-1-tritylpyrazole
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=C(C=N4)Br
Color White
Quantity 1g
Beilstein 550183
Formula Weight 389.3
Physical Form Powder
Percent Purity 95%
Chemical Name or Material 4-Bromo-1-trityl-1H-pyrazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning