Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-1H-imidazole, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2302-25-2
C3H3BrN2
146.97
FHZALEJIENDROK-UHFFFAOYSA-N
4-bromo-1h-imidazole,4-bromoimidazole,1h-imidazole, 4-bromo,4 or 5-bromoimidazole,imidazole, 4-bromo,4-bromo-3h-imidazole,imidazole, 4 or 5-bromo-6ci,7ci,5-bromoimidazole,3kqm,4-bromo-imidazole
96125
5-bromo-1H-imidazole
C1=C(NC=N1)Br
2302-25-2
C3H3BrN2
146.97
FHZALEJIENDROK-UHFFFAOYSA-N
4-bromo-1h-imidazole,4-bromoimidazole,1h-imidazole, 4-bromo,4 or 5-bromoimidazole,imidazole, 4-bromo,4-bromo-3h-imidazole,imidazole, 4 or 5-bromo-6ci,7ci,5-bromoimidazole,3kqm,4-bromo-imidazole
96125
5-bromo-1H-imidazole
C1=C(NC=N1)Br
Melting Point 126.0°C to 132.0°C
Color Beige to Colorless
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 1g
Assay Percent Range 96% min. (HPLC)
Formula Weight 146.97
Physical Form Crystalline Flakes or Powder
Percent Purity 97%
Chemical Name or Material 4-Bromo-1H-imidazole

GHS Signal Word: Danger