Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-1H-indazole, ≥97%

Click to view available options
Quantity:
1g
5g
25g
186407-74-9
C7H5BrN2
197.035
MFCD05664001
KJIODOACRIRBPB-UHFFFAOYSA-N
4-bromoindazole,1h-indazole, 4-bromo,4-bromo 1h indazole,4-bromo 1h indazole,4-bromo-indazole,bromo-1h-indazole,pubchem7834,4-bromo-2h-indazole,acmc-1bzds,4-bromanyl-1h-indazole
22352548
4-bromo-1H-indazole
C1=CC2=C(C=NN2)C(=C1)Br
186407-74-9
C7H5BrN2
197.035
MFCD05664001
KJIODOACRIRBPB-UHFFFAOYSA-N
4-bromoindazole,1h-indazole, 4-bromo,4-bromo 1h indazole,4-bromo 1h indazole,4-bromo-indazole,bromo-1h-indazole,pubchem7834,4-bromo-2h-indazole,acmc-1bzds,4-bromanyl-1h-indazole
22352548
4-bromo-1H-indazole
C1=CC2=C(C=NN2)C(=C1)Br
Quantity 1g
Color White
Melting Point 164°C to 169°C
UN Number UN2811
Beilstein 9757929
Formula Weight 197.04
Physical Form Solid
Percent Purity ≥97%
Chemical Name or Material 4-Bromo-1H-indazole

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

storageNote1 : 2° to 8°C