Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-2,2-difluoro-1,3-benzodioxole, 97%

Click to view available options
Quantity:
1g
5g
144584-66-7
C7H3BrF2O2
237
MFCD01631385
LSZYHXNOLVSZHH-UHFFFAOYSA-N
4-bromo-2,2-difluorobenzo d 1,3 dioxole,4-bromo-2,2-difluoro-2h-1,3-benzodioxole,2,2-difluoro-4-bromo-1,3-benzodioxole,1,3-benzodioxol-4-bromo, 2,2-difluoro,4-bromo-2,2-difluoro-benzo 1,3 dioxole,4-bromo-2,2-difluorobenzo d 1,3-dioxolene,acmc-20a3ue,4-bromo-2,2-difluoro-1,3-benzodioxol,1,3-benzodioxole,4-bromo-2,2-difluoro,4-bromo-2,2-difluoro-1,3-benzodioxolane
2773297
4-bromo-2,2-difluoro-1,3-benzodioxole
C1=CC2=C(C(=C1)Br)OC(O2)(F)F
144584-66-7
C7H3BrF2O2
237
MFCD01631385
LSZYHXNOLVSZHH-UHFFFAOYSA-N
4-bromo-2,2-difluorobenzo d 1,3 dioxole,4-bromo-2,2-difluoro-2h-1,3-benzodioxole,2,2-difluoro-4-bromo-1,3-benzodioxole,1,3-benzodioxol-4-bromo, 2,2-difluoro,4-bromo-2,2-difluoro-benzo 1,3 dioxole,4-bromo-2,2-difluorobenzo d 1,3-dioxolene,acmc-20a3ue,4-bromo-2,2-difluoro-1,3-benzodioxol,1,3-benzodioxole,4-bromo-2,2-difluoro,4-bromo-2,2-difluoro-1,3-benzodioxolane
2773297
4-bromo-2,2-difluoro-1,3-benzodioxole
C1=CC2=C(C(=C1)Br)OC(O2)(F)F
Applications
4-Bromo-2,2-difluoro-1,3-benzodioxole is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
It is unstable under normal temperature. storage temperature is 2 - 4°C. Incompatible material is oxidizing agents.
Quantity 1g
Formula Weight 237
Percent Purity 97%
Chemical Name or Material 4-Bromo-2,2-difluoro-1,3-benzodioxole

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning