Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-2-(2-nitro-1-propenyl)thiophene, 97%

Click to view available options
Quantity:
1g
5g
175205-19-3
C7H6BrNO2S
248.094
MFCD00052971
DEFWRSIMNNRSKK-DJWKRKHSSA-N
4-bromo-2-2-nitroprop-1-enyl thiophene,4-bromo-2-2-nitro-1-propenyl thiophene,4-bromo-2-z-2-nitroprop-1-enyl thiophene,2-1z-2-nitroprop-1-enyl-4-bromothiophene
5906915
4-bromo-2-[(Z)-2-nitroprop-1-enyl]thiophene
CC(=CC1=CC(=CS1)Br)[N+](=O)[O-]
175205-19-3
C7H6BrNO2S
248.094
MFCD00052971
DEFWRSIMNNRSKK-DJWKRKHSSA-N
4-bromo-2-2-nitroprop-1-enyl thiophene,4-bromo-2-2-nitro-1-propenyl thiophene,4-bromo-2-z-2-nitroprop-1-enyl thiophene,2-1z-2-nitroprop-1-enyl-4-bromothiophene
5906915
4-bromo-2-[(Z)-2-nitroprop-1-enyl]thiophene
CC(=CC1=CC(=CS1)Br)[N+](=O)[O-]
Quantity 5g
Melting Point 88°C to 90°C
Formula Weight 248.09
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-(2-nitro-1-propenyl)thiophene

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger