Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2,5-difluoroanisole, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
202865-60-9
C7H5BrF2O
223.017
MFCD00143258
OOTXNQBDWFJMNN-UHFFFAOYSA-N
4-bromo-2,5-difluoroanisole,1-bromo-2,5-difluoro-4-methoxy-benzene,timtec-bb sbb037912,pubchem2653,acmc-1cm1k,benzene,1-bromo-2,5-difluoro-4-methoxy,benzene, 1-bromo-2,5-difluoro-4-methoxy
2724983
1-bromo-2,5-difluoro-4-methoxybenzene
COC1=CC(=C(C=C1F)Br)F
202865-60-9
C7H5BrF2O
223.017
MFCD00143258
OOTXNQBDWFJMNN-UHFFFAOYSA-N
4-bromo-2,5-difluoroanisole,1-bromo-2,5-difluoro-4-methoxy-benzene,timtec-bb sbb037912,pubchem2653,acmc-1cm1k,benzene,1-bromo-2,5-difluoro-4-methoxy,benzene, 1-bromo-2,5-difluoro-4-methoxy
2724983
1-bromo-2,5-difluoro-4-methoxybenzene
COC1=CC(=C(C=C1F)Br)F
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
Color White
Melting Point 59°C to 63°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 223.02
Physical Form Crystals
Percent Purity ≥97.5%
Chemical Name or Material 4-Bromo-2, 5-difluoroanisole, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.