Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-2,5-difluorobenzenesulfonyl chloride, 97%

Click to view available options
Quantity:
1g
5g
207974-14-9
C6H2BrClF2O2S
291.491
MFCD00042181
SBMKFWMFNIEPDN-UHFFFAOYSA-N
4-bromo-2,5-difluorobenzene-1-sulfonylchloride,4-bromo-2,5-difluorobenzenesulphonyl chloride,4-bromo-2,5-difluorobenzene-1-sulfonyl chloride,4-bromo-2,5-difluorophenyl chlorosulfone,4-bromo-2,5-difluorobenzenesulphonylchloride,benzenesulfonyl chloride, 4-bromo-2,5-difluoro,4-bromo-3,6-difluoro-benzenesulfonylchloride,pubchem5058,acmc-209feg,4-bromo-2,5-difluorobenzenesulfonylchloride
574590
4-bromo-2,5-difluorobenzenesulfonyl chloride
C1=C(C(=CC(=C1S(=O)(=O)Cl)F)Br)F
207974-14-9
C6H2BrClF2O2S
291.491
MFCD00042181
SBMKFWMFNIEPDN-UHFFFAOYSA-N
4-bromo-2,5-difluorobenzene-1-sulfonylchloride,4-bromo-2,5-difluorobenzenesulphonyl chloride,4-bromo-2,5-difluorobenzene-1-sulfonyl chloride,4-bromo-2,5-difluorophenyl chlorosulfone,4-bromo-2,5-difluorobenzenesulphonylchloride,benzenesulfonyl chloride, 4-bromo-2,5-difluoro,4-bromo-3,6-difluoro-benzenesulfonylchloride,pubchem5058,acmc-209feg,4-bromo-2,5-difluorobenzenesulfonylchloride
574590
4-bromo-2,5-difluorobenzenesulfonyl chloride
C1=C(C(=CC(=C1S(=O)(=O)Cl)F)Br)F
Sensitivity Moisture sensitive
Boiling Point 76°C (0.01mmHg)
Flash Point >110°C (230°F)
Quantity 1g
Melting Point 34°C to 36°C
UN Number UN3261
Formula Weight 291.5
Percent Purity 97%
Chemical Name or Material 4-Bromo-2,5-difluorobenzenesulfonyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen