Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-2,5-dimethoxybenzaldehyde, 98+%

Click to view available options
Quantity:
5g
25g
31558-41-5
C9H9BrO3
245.072
MFCD00156964
BIFWGDWGCZLCHF-UHFFFAOYSA-N
2,5-dimethoxy-4-bromobenzaldehyde,pubchem2068,acmc-1aiq9,cbmicro_047429,cambridge id 6238031,4-bromo-2,5-dimethoxy benzaldehyde,4-bromo-2,5-dimethoxy-benzaldehyde,4-bromanyl-2,5-dimethoxy-benzaldehyde,benzaldehyde, 4-bromo-2,5-dimethoxy
777997
4-bromo-2,5-dimethoxybenzaldehyde
COC1=CC(=C(C=C1C=O)OC)Br
31558-41-5
C9H9BrO3
245.072
MFCD00156964
BIFWGDWGCZLCHF-UHFFFAOYSA-N
2,5-dimethoxy-4-bromobenzaldehyde,pubchem2068,acmc-1aiq9,cbmicro_047429,cambridge id 6238031,4-bromo-2,5-dimethoxy benzaldehyde,4-bromo-2,5-dimethoxy-benzaldehyde,4-bromanyl-2,5-dimethoxy-benzaldehyde,benzaldehyde, 4-bromo-2,5-dimethoxy
777997
4-bromo-2,5-dimethoxybenzaldehyde
COC1=CC(=C(C=C1C=O)OC)Br
Applications
4-Bromo-2,5-dimethoxybenzene is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Incompatible with oxidizing agents.
Melting Point 132°C to 135°C
Quantity 25g
Formula Weight 245.08
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-2,5-dimethoxybenzaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning