Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2,6-difluoroaniline, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
67567-26-4
C6H4BrF2N
208.01
BFQSQUAVMNHOEF-UHFFFAOYSA-N
2,6-difluoro-4-bromoaniline,4-bromo-2,6-difluorobenzenamine,benzenamine, 4-bromo-2,6-difluoro,4-bromo-2,6-difluoro-phenylamine,4-bromo-2,6-difluoro-aniline,4-bromo-2,6-difluorophenylamine,buttpark 22\07-66,4-bromo-2,6-difluorophenyl amine,pubchem2929,acmc-1begp
610191
4-bromo-2,6-difluoroaniline
C1=C(C=C(C(=C1F)N)F)Br
67567-26-4
C6H4BrF2N
208.01
BFQSQUAVMNHOEF-UHFFFAOYSA-N
2,6-difluoro-4-bromoaniline,4-bromo-2,6-difluorobenzenamine,benzenamine, 4-bromo-2,6-difluoro,4-bromo-2,6-difluoro-phenylamine,4-bromo-2,6-difluoro-aniline,4-bromo-2,6-difluorophenylamine,buttpark 22\07-66,4-bromo-2,6-difluorophenyl amine,pubchem2929,acmc-1begp
610191
4-bromo-2,6-difluoroaniline
C1=C(C=C(C(=C1F)N)F)Br
Melting Point 67°C to 68°C
Color Brown
Infrared Spectrum Authentic
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 208.01
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-Bromo-2, 6-difluoroaniline
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning