Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-2,6-difluorobenzeneboronic acid, 98%

Click to view available options
Quantity:
1g
5g
352535-81-0
C6H4BBrF2O2
236.807
MFCD03095369
QHYNAULNEIXXPC-UHFFFAOYSA-N
4-bromo-2,6-difluorophenyl boronic acid,2,6-difluoro-4-bromophenylboronic acid,4-bromo-2,6-difluorobenzeneboronic acid,boronic acid, b-4-bromo-2,6-difluorophenyl,4-bromo-2 6-difluorophenylboronic acid,boronic acid,b-4-bromo-2,6-difluorophenyl,pubchem1757,acmc-209if9,4-bromo-2,6-difluorophenylboronicacid,4-bromanyl-2,6-bis fluoranyl phenyl boronic acid
4125779
(4-bromo-2,6-difluorophenyl)boronic acid
B(C1=C(C=C(C=C1F)Br)F)(O)O
352535-81-0
C6H4BBrF2O2
236.807
MFCD03095369
QHYNAULNEIXXPC-UHFFFAOYSA-N
4-bromo-2,6-difluorophenyl boronic acid,2,6-difluoro-4-bromophenylboronic acid,4-bromo-2,6-difluorobenzeneboronic acid,boronic acid, b-4-bromo-2,6-difluorophenyl,4-bromo-2 6-difluorophenylboronic acid,boronic acid,b-4-bromo-2,6-difluorophenyl,pubchem1757,acmc-209if9,4-bromo-2,6-difluorophenylboronicacid,4-bromanyl-2,6-bis fluoranyl phenyl boronic acid
4125779
(4-bromo-2,6-difluorophenyl)boronic acid
B(C1=C(C=C(C=C1F)Br)F)(O)O
Quantity 1g
Melting Point 155°C to 160°C (literature)
Formula Weight 236.81
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 4-Bromo-2,6-difluorobenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning