Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-2,6-difluorobenzotrifluoride, 98%

Click to view available options
Quantity:
250mg
1g
5g
156243-64-0
C7H2BrF5
260.989
MFCD01631616
QPJKIRNIIXIPIE-UHFFFAOYSA-N
3,5-difluoro-4-trifluoromethyl bromobenzene,5-bromo-1,3-difluoro-2-trifluoromethyl benzene,4-bromo-2,6-difluorobenzotrifluoride,benzene, 5-bromo-1,3-difluoro-2-trifluoromethyl,acmc-1cioh,intermediates-zcf02186,2,6-difluoro-4-bromobenzotrifluoride,1-bromo-3,5-difluoro-4-trifluoromethylbenzene,3,5-difluoro-4-trifluoromethylbromobenzene,benzene,5-bromo-1,3-difluoro-2-trifluoromethyl
23450919
5-bromo-1,3-difluoro-2-(trifluoromethyl)benzene
C1=C(C=C(C(=C1F)C(F)(F)F)F)Br
156243-64-0
C7H2BrF5
260.989
MFCD01631616
QPJKIRNIIXIPIE-UHFFFAOYSA-N
3,5-difluoro-4-trifluoromethyl bromobenzene,5-bromo-1,3-difluoro-2-trifluoromethyl benzene,4-bromo-2,6-difluorobenzotrifluoride,benzene, 5-bromo-1,3-difluoro-2-trifluoromethyl,acmc-1cioh,intermediates-zcf02186,2,6-difluoro-4-bromobenzotrifluoride,1-bromo-3,5-difluoro-4-trifluoromethylbenzene,3,5-difluoro-4-trifluoromethylbromobenzene,benzene,5-bromo-1,3-difluoro-2-trifluoromethyl
23450919
5-bromo-1,3-difluoro-2-(trifluoromethyl)benzene
C1=C(C=C(C(=C1F)C(F)(F)F)F)Br
Applications
It is employed in the lewis acid-promoted synthesis of unsymmetrical and highly functionalized carbazoles and dibenzofurans from biaryl triazenes.

Solubility
Slightly soluble in water.

Notes
Store away from bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat.
Quantity 250mg
Color Colorless
Beilstein 13252396
Formula Weight 260.99
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-Bromo-2,6-difluorobenzotrifluoride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning