Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-2,6-difluorophenylacetic acid, 96%

Click to view available options
Quantity:
250mg
1g
537033-54-8
C8H5BrF2O2
251.027
MFCD03094466
GBFFYYLXGIHKLU-UHFFFAOYSA-N
4-bromo-2,6-difluorophenylacetic acid,2-4-bromo-2,6-difluorophenyl acetic acid,4-bromo-2,6-difluorophenyl acetic acid,2-4-bromo-2,6-difluoro-phenyl acetic acid,acmc-209uz8,4-bromo-2,6-difluorophenylaceticacid,4-bromo-2,6-difluorobenzeneacetic acid,4-bromo-2,6-difluoro-phenyl-acetic acid,4-bromo-2,6-difluorophenylacetic acid,benzeneacetic acid, 4-bromo-2,6-difluoro
2773309
2-(4-bromo-2,6-difluorophenyl)acetic acid
C1=C(C=C(C(=C1F)CC(=O)O)F)Br
537033-54-8
C8H5BrF2O2
251.027
MFCD03094466
GBFFYYLXGIHKLU-UHFFFAOYSA-N
4-bromo-2,6-difluorophenylacetic acid,2-4-bromo-2,6-difluorophenyl acetic acid,4-bromo-2,6-difluorophenyl acetic acid,2-4-bromo-2,6-difluoro-phenyl acetic acid,acmc-209uz8,4-bromo-2,6-difluorophenylaceticacid,4-bromo-2,6-difluorobenzeneacetic acid,4-bromo-2,6-difluoro-phenyl-acetic acid,4-bromo-2,6-difluorophenylacetic acid,benzeneacetic acid, 4-bromo-2,6-difluoro
2773309
2-(4-bromo-2,6-difluorophenyl)acetic acid
C1=C(C=C(C(=C1F)CC(=O)O)F)Br
Quantity 250mg
Density 1.77g/mL
Formula Weight 251.02
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 4-Bromo-2,6-difluorophenylacetic acid