Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-2,6-dimethylaniline, 98%

Manufacturer:  Alfa Aesar™ A1071230

Quantity 250g

 View more versions of this product

Catalog No. AAA1071230


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

24596-19-8
200.08
UN2811
4-Bromo-2,6-xylidine
C8H10BrN
MFCD00007826
1936300
4-Bromo-2,6-dimethylaniline

Specifications

24596-19-8
200.08
UN2811
1936300
120°C (0.2mmHg)
>110°C (230°F)
98%
C8H10BrN
MFCD00007826
47° to 52°C
4-Bromo-2,6-xylidine
250g
4-Bromo-2,6-dimethylaniline

Safety and Handling

SDS & Safety Documents

GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit