Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-2,6-dimethylanisole, 99%

Click to view available options
Quantity:
5g
25g
100g
14804-38-7
C9H11BrO
215.09
MFCD00143259
MMARFGDTMJBIBK-UHFFFAOYSA-N
4-bromo-2,6-dimethylanisole,5-bromo-2-methoxy-m-xylene,4-brom-2,6-dimethylanisole,1-bromo-3,5-dimethyl-4-methoxybenzene,2,6-dimethyl-4-bromoanisole,benzene, 5-bromo-2-methoxy-1,3-dimethyl,timtec-bb sbb005776,attercop-chm at108629,4-bromo-2,6-dimethyl-methoxybenzene,pubchem2654
278654
5-bromo-2-methoxy-1,3-dimethylbenzene
CC1=CC(=CC(=C1OC)C)Br
14804-38-7
C9H11BrO
215.09
MFCD00143259
MMARFGDTMJBIBK-UHFFFAOYSA-N
4-bromo-2,6-dimethylanisole,5-bromo-2-methoxy-m-xylene,4-brom-2,6-dimethylanisole,1-bromo-3,5-dimethyl-4-methoxybenzene,2,6-dimethyl-4-bromoanisole,benzene, 5-bromo-2-methoxy-1,3-dimethyl,timtec-bb sbb005776,attercop-chm at108629,4-bromo-2,6-dimethyl-methoxybenzene,pubchem2654
278654
5-bromo-2-methoxy-1,3-dimethylbenzene
CC1=CC(=CC(=C1OC)C)Br
Flash Point >110°C (230°F)
Quantity 100g
Boiling Point 117°C to 120°C (13mmHg)
Beilstein 1940290
Formula Weight 215.09
Percent Purity 99%
Chemical Name or Material 4-Bromo-2,6-dimethylanisole

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning