Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-2-chloro-6-fluorophenol, 97+%

Click to view available options
Quantity:
1g
5g
161045-79-0
C6H3BrClFO
225.441
MFCD01631476
QEYONPKSDTUPAX-UHFFFAOYSA-N
4-bromo-2-chloro-6-fluoro-phenol,phenol,4-bromo-2-chloro-6-fluoro,pubchem2841,acmc-209dlz,intermediates-zcf02212,3-bromo-2-chloro-6-fluoro-phenol,phenol, 4-bromo-2-chloro-6-fluoro,4-bromo-2-chloro-6-fluorophenol
2773270
4-bromo-2-chloro-6-fluorophenol
C1=C(C=C(C(=C1Cl)O)F)Br
161045-79-0
C6H3BrClFO
225.441
MFCD01631476
QEYONPKSDTUPAX-UHFFFAOYSA-N
4-bromo-2-chloro-6-fluoro-phenol,phenol,4-bromo-2-chloro-6-fluoro,pubchem2841,acmc-209dlz,intermediates-zcf02212,3-bromo-2-chloro-6-fluoro-phenol,phenol, 4-bromo-2-chloro-6-fluoro,4-bromo-2-chloro-6-fluorophenol
2773270
4-bromo-2-chloro-6-fluorophenol
C1=C(C=C(C(=C1Cl)O)F)Br
Quantity 1g
Formula Weight 255.45
Percent Purity ≥97%
Chemical Name or Material 4-Bromo-2-chloro-6-fluorophenol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning