Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-2-chlorobenzenesulfonyl chloride, 96%

Click to view available options
Quantity:
5g
25g
100g
351003-52-6
C6H3BrCl2O2S
289.952
MFCD03094630
OECYEHWJPRPCOH-UHFFFAOYSA-N
4-bromo-2-chlorobenzene-1-sulfonyl chloride,2-chloro-4-bromo-benzenesulfonyl chloride,4-bromo-2-chlorobenzenesulphonyl chloride,4-bromo-2-chlorobenzenesulfonylchloride,benzenesulfonyl chloride, 4-bromo-2-chloro,4-bromo-2-chlorophenylsulfonyl chloride,pubchem11732,4-bromo-2-chloro-benzenesulfonyl chloride,acmc-1ag54,ksc221s9p
2734766
4-bromo-2-chlorobenzenesulfonyl chloride
C1=CC(=C(C=C1Br)Cl)S(=O)(=O)Cl
351003-52-6
C6H3BrCl2O2S
289.952
MFCD03094630
OECYEHWJPRPCOH-UHFFFAOYSA-N
4-bromo-2-chlorobenzene-1-sulfonyl chloride,2-chloro-4-bromo-benzenesulfonyl chloride,4-bromo-2-chlorobenzenesulphonyl chloride,4-bromo-2-chlorobenzenesulfonylchloride,benzenesulfonyl chloride, 4-bromo-2-chloro,4-bromo-2-chlorophenylsulfonyl chloride,pubchem11732,4-bromo-2-chloro-benzenesulfonyl chloride,acmc-1ag54,ksc221s9p
2734766
4-bromo-2-chlorobenzenesulfonyl chloride
C1=CC(=C(C=C1Br)Cl)S(=O)(=O)Cl
Melting Point 76°C to 81°C
Quantity 5g
UN Number UN3261
Formula Weight 289.96
Percent Purity 96%
Chemical Name or Material 4-Bromo-2-chlorobenzenesulfonyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.