Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-2-chlorophenol, 98%

Click to view available options
Quantity:
10g
50g
250g
3964-56-5
C6H4BrClO
207.451
MFCD00002166
VIBJPUXLAKVICD-UHFFFAOYSA-N
2-chloro-4-bromophenol,phenol, 4-bromo-2-chloro,phenol, 4-bromochloro,unii-994sj34d8p,4-bromo-2-chloro-phenol,attercop-chm at106179,4-bromo-2-chlorophenol 25gr,4-bromo-chlorophenol,pubchem3596,acmc-209j6p
19859
CHEBI:38852
4-bromo-2-chlorophenol
C1=CC(=C(C=C1Br)Cl)O
3964-56-5
C6H4BrClO
207.451
MFCD00002166
VIBJPUXLAKVICD-UHFFFAOYSA-N
2-chloro-4-bromophenol,phenol, 4-bromo-2-chloro,phenol, 4-bromochloro,unii-994sj34d8p,4-bromo-2-chloro-phenol,attercop-chm at106179,4-bromo-2-chlorophenol 25gr,4-bromo-chlorophenol,pubchem3596,acmc-209j6p
19859
CHEBI:38852
4-bromo-2-chlorophenol
C1=CC(=C(C=C1Br)Cl)O
Melting Point 47°C to 50°C
Flash Point >110°C (230°F)
Quantity 250g
Boiling Point 232°C to 235°C
Odor Phenol-like
Beilstein 2042867
Formula Weight 207.46
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-chlorophenol

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning