Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 4-Bromo-2-chlorophenol, 98%
Click to view available options
Quantity:
10g
50g
250g
Specifications
Odor | Phenol-like |
Boiling Point | 232°C to 235°C |
Flash Point | >110°C (230°F) |
Melting Point | 47°C to 50°C |
Quantity | 10g |
Beilstein | 2042867 |
Formula Weight | 207.46 |
Percent Purity | 98% |
Chemical Name or Material | 4-Bromo-2-chlorophenol |
Safety and Handling
GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 223-572-5
TSCA : No
Recommended Storage : Ambient temperatures