Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-chloropyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
73583-37-6
C5H3BrClN
192.44
MFCD03840756
ONHMWUXYIFULDO-UHFFFAOYSA-N
2-chloro-4-bromopyridine,pyridine, 4-bromo-2-chloro,4-bromo-2-chloro-pyridine,pubchem1171,acmc-1bjmw,ksc381c4t,tpc-py061,4-bromo-2-chloropyridine,abbypharma ap-18-5387,4-bromo-2-chloropyridine 5g
3847770
4-bromo-2-chloropyridine
C1=CN=C(C=C1Br)Cl
73583-37-6
C5H3BrClN
192.44
MFCD03840756
ONHMWUXYIFULDO-UHFFFAOYSA-N
2-chloro-4-bromopyridine,pyridine, 4-bromo-2-chloro,4-bromo-2-chloro-pyridine,pubchem1171,acmc-1bjmw,ksc381c4t,tpc-py061,4-bromo-2-chloropyridine,abbypharma ap-18-5387,4-bromo-2-chloropyridine 5g
3847770
4-bromo-2-chloropyridine
C1=CN=C(C=C1Br)Cl
Specific Gravity 1.734
Flash Point 104°C
Color Yellow
Infrared Spectrum Authentic
Density 1.7340g/mL
Packaging Glass bottle
Quantity 25g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 192.44
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-chloropyridine
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning