Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-2-chloropyrimidine, 98%

Click to view available options
Quantity:
250mg
1g
885702-34-1
C4H2BrClN2
193.428
MFCD08272196
FBEBVAQOMVWORE-UHFFFAOYSA-N
pyrimidine, 4-bromo-2-chloro-9ci,pyrimidine, 4-bromo-2-chloro,2-chloro-4-bromopyrimidine
45117394
4-bromo-2-chloropyrimidine
C1=CN=C(N=C1Br)Cl
885702-34-1
C4H2BrClN2
193.428
MFCD08272196
FBEBVAQOMVWORE-UHFFFAOYSA-N
pyrimidine, 4-bromo-2-chloro-9ci,pyrimidine, 4-bromo-2-chloro,2-chloro-4-bromopyrimidine
45117394
4-bromo-2-chloropyrimidine
C1=CN=C(N=C1Br)Cl
Applications
Used as an primary and secondary intermediate for pharmaceutical, organic synthesis.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. With adequate ventilation. Store away from oxidizing agent.
Quantity 250mg
Beilstein 10334333
Formula Weight 193.43
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-chloropyrimidine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning