Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-2-chlorotoluene, 98%

Click to view available options
Quantity:
25g
100g
89794-02-5
C7H6BrCl
205.479
MFCD00060651
LIFMTDJMLRECMX-UHFFFAOYSA-N
4-bromo-2-chlorotoluene,2-chloro-4-bromotoluene,4-bromo-2-chloro toluene,4-bromo-2-chloro-1-methyl-benzene,benzene, 4-bromo-2-chloro-1-methyl,4-bromo-chlorotoluene,pubchem3603,4-bromo-2chlorotoluene,2-chloro 4-bromo toluene,acmc-209r2x
2735554
4-bromo-2-chloro-1-methylbenzene
CC1=C(C=C(C=C1)Br)Cl
89794-02-5
C7H6BrCl
205.479
MFCD00060651
LIFMTDJMLRECMX-UHFFFAOYSA-N
4-bromo-2-chlorotoluene,2-chloro-4-bromotoluene,4-bromo-2-chloro toluene,4-bromo-2-chloro-1-methyl-benzene,benzene, 4-bromo-2-chloro-1-methyl,4-bromo-chlorotoluene,pubchem3603,4-bromo-2chlorotoluene,2-chloro 4-bromo toluene,acmc-209r2x
2735554
4-bromo-2-chloro-1-methylbenzene
CC1=C(C=C(C=C1)Br)Cl
Boiling Point 212°C
Flash Point 104°C (219°F)
Quantity 100g
Beilstein 3234853
Formula Weight 205.48
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-chlorotoluene

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning