Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4'-Bromo-2'-fluoroacetanilide, 98%

Click to view available options
Quantity:
10g
50g
250g
326-66-9
C8H7BrFNO
232.052
MFCD00042588
BCYGKMDWQBWUSC-UHFFFAOYSA-N
n-4-bromo-2-fluorophenyl acetamide,4'-bromo-2'-fluoroacetanilide,4-bromo-2-fluoroacetanilide,acetamide, n-4-bromo-2-fluorophenyl,pubchem14886,acmc-1cooc,ksc497m2b,timtec-bb sbb009911,attercop-chm at130693
67593
N-(4-bromo-2-fluorophenyl)acetamide
CC(=O)NC1=C(C=C(C=C1)Br)F
326-66-9
C8H7BrFNO
232.052
MFCD00042588
BCYGKMDWQBWUSC-UHFFFAOYSA-N
n-4-bromo-2-fluorophenyl acetamide,4'-bromo-2'-fluoroacetanilide,4-bromo-2-fluoroacetanilide,acetamide, n-4-bromo-2-fluorophenyl,pubchem14886,acmc-1cooc,ksc497m2b,timtec-bb sbb009911,attercop-chm at130693
67593
N-(4-bromo-2-fluorophenyl)acetamide
CC(=O)NC1=C(C=C(C=C1)Br)F
Quantity 250g
Melting Point 154°C to 158°C
Beilstein 3250459
Formula Weight 232.05
Percent Purity 98%
Chemical Name or Material 4'-Bromo-2'-fluoroacetanilide

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 206-314-6

TSCA : No

Recommended Storage : Ambient temperatures