Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4'-Bromo-2'-fluoroacetophenone, 96%

Click to view available options
Quantity:
250mg
1g
5g
625446-22-2
C8H6BrFO
217.037
MFCD03411551
ASKFCSCYGAFWAB-UHFFFAOYSA-N
1-4-bromo-2-fluorophenyl ethanone,4-bromo-2-fluoroacetophenone,4'-bromo-2'-fluoroacetophenone,1-4-bromo-2-fluorophenyl ethan-1-one,1-acetyl-4-bromo-2-fluorobenzene,ethanone, 1-4-bromo-2-fluorophenyl,2-fluoro-4-bromoacetophenone,1-4-bromo-2-fluoro-phenyl-ethanone,acmc-209n4s,ksc352o4j
21938581
1-(4-bromo-2-fluorophenyl)ethanone
CC(=O)C1=C(C=C(C=C1)Br)F
625446-22-2
C8H6BrFO
217.037
MFCD03411551
ASKFCSCYGAFWAB-UHFFFAOYSA-N
1-4-bromo-2-fluorophenyl ethanone,4-bromo-2-fluoroacetophenone,4'-bromo-2'-fluoroacetophenone,1-4-bromo-2-fluorophenyl ethan-1-one,1-acetyl-4-bromo-2-fluorobenzene,ethanone, 1-4-bromo-2-fluorophenyl,2-fluoro-4-bromoacetophenone,1-4-bromo-2-fluoro-phenyl-ethanone,acmc-209n4s,ksc352o4j
21938581
1-(4-bromo-2-fluorophenyl)ethanone
CC(=O)C1=C(C=C(C=C1)Br)F
Quantity 5g
Formula Weight 217.04
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 4'-Bromo-2'-fluoroacetophenone

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures