Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-fluoroaniline 99%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
367-24-8
C6H5BrFN
190.01
MFCD00010221
GZRMNMGWNKSANY-UHFFFAOYSA-N
2-fluoro-4-bromoaniline,benzenamine, 4-bromo-2-fluoro,4-bromo-2-fluoro-phenylamine,4-bromo-2-fluoro-aniline,4-bromo-2-fluorobenzenamine,4-bromo-2-fluoro aniline,4-bromo-2-fluorophenylamine,4-brom-2-fluoranilin,zlchem 545,pubchem1525
123050
4-bromo-2-fluoroaniline
C1=CC(=C(C=C1Br)F)N
367-24-8
C6H5BrFN
190.01
MFCD00010221
GZRMNMGWNKSANY-UHFFFAOYSA-N
2-fluoro-4-bromoaniline,benzenamine, 4-bromo-2-fluoro,4-bromo-2-fluoro-phenylamine,4-bromo-2-fluoro-aniline,4-bromo-2-fluorobenzenamine,4-bromo-2-fluoro aniline,4-bromo-2-fluorophenylamine,4-brom-2-fluoranilin,zlchem 545,pubchem1525
123050
4-bromo-2-fluoroaniline
C1=CC(=C(C=C1Br)F)N
Boiling Point 103°C to 108°C (13.0mmHg)
Melting Point 39°C to 42°C
Flash Point 104°C
Quantity 10g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Beige to Brown-Gray or Gray
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula BrC6H3(F)NH2
Formula Weight 190.01
Physical Form Crystalline Mass, Powder, Crystals and/or Chunks
Percent Purity ≥98.5%
Chemical Name or Material 4-Bromo-2-fluoroaniline, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-685-4