Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-2-fluorobenzenesulfonyl chloride, 98%

Click to view available options
Quantity:
1g
5g
25g
216159-03-4
C6H3BrClFO2S
273.5
MFCD01090993
XNYBZLRIUHNRQY-UHFFFAOYSA-N
4-bromo-2-fluorobenzene-1-sulfonyl chloride,2-fluoro-4-bromobenzenesulfonyl chloride,4-bromo-2-fluorobenzenesulphonyl chloride,4-bromo-2-fluoro-benzenesulfonyl chloride,buttpark 147\11-41,4-bromo-2-fluorophenyl chlorosulfone,benzenesulfonyl chloride, 4-bromo-2-fluoro,pubchem5065,acmc-209via,ksc201o8l
2736310
4-bromo-2-fluorobenzenesulfonyl chloride
C1=CC(=C(C=C1Br)F)S(=O)(=O)Cl
216159-03-4
C6H3BrClFO2S
273.5
MFCD01090993
XNYBZLRIUHNRQY-UHFFFAOYSA-N
4-bromo-2-fluorobenzene-1-sulfonyl chloride,2-fluoro-4-bromobenzenesulfonyl chloride,4-bromo-2-fluorobenzenesulphonyl chloride,4-bromo-2-fluoro-benzenesulfonyl chloride,buttpark 147\11-41,4-bromo-2-fluorophenyl chlorosulfone,benzenesulfonyl chloride, 4-bromo-2-fluoro,pubchem5065,acmc-209via,ksc201o8l
2736310
4-bromo-2-fluorobenzenesulfonyl chloride
C1=CC(=C(C=C1Br)F)S(=O)(=O)Cl
Melting Point 63°C to 66°C
Quantity 5g
Boiling Point 84°C (0.5mmHg)
UN Number UN3261
Formula Weight 273.51
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-fluorobenzenesulfonyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.