Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-fluorobenzoic acid, 98%, Acros Organics

Click to view available options
Quantity:
5g
112704-79-7
C7H4BrFO2
219.009
MFCD00143263
ZQQSRVPOAHYHEL-UHFFFAOYSA-N
2-fluoro-4-bromobenzoic acid,4-bromo-2-fluoro-benzoic acid,4-bromo-2-fluorobenzoicacid,benzoic acid, 4-bromo-2-fluoro,pubchem1313,enzalutamide impurity a,acmc-1btmf,2-fluro-4-bromobenzoic acid,ksc174m4p,rarechem al bo 0754
302681
4-bromo-2-fluorobenzoic acid
C1=CC(=C(C=C1Br)F)C(=O)O
112704-79-7
C7H4BrFO2
219.009
MFCD00143263
ZQQSRVPOAHYHEL-UHFFFAOYSA-N
2-fluoro-4-bromobenzoic acid,4-bromo-2-fluoro-benzoic acid,4-bromo-2-fluorobenzoicacid,benzoic acid, 4-bromo-2-fluoro,pubchem1313,enzalutamide impurity a,acmc-1btmf,2-fluro-4-bromobenzoic acid,ksc174m4p,rarechem al bo 0754
302681
4-bromo-2-fluorobenzoic acid
C1=CC(=C(C=C1Br)F)C(=O)O
Quantity 5g
Color White to Yellow
Infrared Spectrum Authentic
Melting Point 208°C to 213°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula BrC6H3(F)COOH
Formula Weight 219.01
Physical Form Crystalline Powder or Crystals
Percent Purity ≥97.5%
Chemical Name or Material 4-Bromo-2-fluorobenzoic acid, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.