Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-fluorobenzyl alcohol, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
188582-62-9
C7H6BrFO
205.026
MFCD00143265
BWBJZMQPVBWEJU-UHFFFAOYSA-N
4-bromo-2-fluorobenzyl alcohol,4-bromo-2-fluorophenyl methanol,4-bromo-2-fluorobenzylalcohol,benzenemethanol, 4-bromo-2-fluoro,rarechem al bd 0386,timtec-bb sbb005826,4-bromo-2-fluorophenyl methan-1-ol,2-fluoro-4-bromobenzyl alcohol,pubchem3223,acmc-209erg
2734027
(4-bromo-2-fluorophenyl)methanol
C1=CC(=C(C=C1Br)F)CO
188582-62-9
C7H6BrFO
205.026
MFCD00143265
BWBJZMQPVBWEJU-UHFFFAOYSA-N
4-bromo-2-fluorobenzyl alcohol,4-bromo-2-fluorophenyl methanol,4-bromo-2-fluorobenzylalcohol,benzenemethanol, 4-bromo-2-fluoro,rarechem al bd 0386,timtec-bb sbb005826,4-bromo-2-fluorophenyl methan-1-ol,2-fluoro-4-bromobenzyl alcohol,pubchem3223,acmc-209erg
2734027
(4-bromo-2-fluorophenyl)methanol
C1=CC(=C(C=C1Br)F)CO
Quantity 1g
Melting Point 31°C to 35°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color White to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 205.03
Physical Form Crystalline Powder or Chunks
Percent Purity ≥97.5%
Chemical Name or Material 4-Bromo-2-fluorobenzyl alcohol, 98%
Show More Show Less