Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-fluorobenzyl bromide, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Plastic bottle
76283-09-5
C7H5Br2F
267.92
MFCD00055467
XMHNLZXYPAULDF-UHFFFAOYSA-N
4-bromo-2-fluorobenzyl bromide,4-bromo-1-bromomethyl-2-fluorobenzene,2-fluoro-4-bromobenzyl bromide,4-bromo-2-fluorobenzylbromide,2-fluoro-4-bromo-benzylbromide,alpha,4-dibromo-2-fluorotoluene,unii-0z26xyc1jy,4,alpha-dibromo-2-fluorotoluene,4-bromo-1-bromomethyl-2-fluoro-benzene,0z26xyc1jy
2733660
4-bromo-1-(bromomethyl)-2-fluorobenzene
C1=CC(=C(C=C1Br)F)CBr
76283-09-5
C7H5Br2F
267.92
MFCD00055467
XMHNLZXYPAULDF-UHFFFAOYSA-N
4-bromo-2-fluorobenzyl bromide,4-bromo-1-bromomethyl-2-fluorobenzene,2-fluoro-4-bromobenzyl bromide,4-bromo-2-fluorobenzylbromide,2-fluoro-4-bromo-benzylbromide,alpha,4-dibromo-2-fluorotoluene,unii-0z26xyc1jy,4,alpha-dibromo-2-fluorotoluene,4-bromo-1-bromomethyl-2-fluoro-benzene,0z26xyc1jy
2733660
4-bromo-1-(bromomethyl)-2-fluorobenzene
C1=CC(=C(C=C1Br)F)CBr
Melting Point >30.0°C
Color White
Quantity 5g
Packaging Plastic bottle
Infrared Spectrum Authentic
Boiling Point 126.0°C (19.0 mmHg)
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 267.92
Physical Form Low Melting Solid
Percent Purity 99%
Chemical Name or Material 4-Bromo-2-fluorobenzyl bromide
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 278-412-7