Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-bromo-2-fluorobenzyl Chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
85510-82-3
C7H5BrClF
223.47
MFCD02094036
UDKQGFMDBMYVHI-UHFFFAOYSA-N
4-bromo-2-fluorobenzyl chloride,4-bromo-1-chloromethyl-2-fluorobenzene,4-bromo-2-fluorobenzylchloride,4-bromo-1-chloromethyl-2-fluoro-benzene,acmc-209q6a,2-fluoro-4-bromo-chloromethylbenzene
2773361
4-bromo-1-(chloromethyl)-2-fluorobenzene
C1=CC(=C(C=C1Br)F)CCl
85510-82-3
C7H5BrClF
223.47
MFCD02094036
UDKQGFMDBMYVHI-UHFFFAOYSA-N
4-bromo-2-fluorobenzyl chloride,4-bromo-1-chloromethyl-2-fluorobenzene,4-bromo-2-fluorobenzylchloride,4-bromo-1-chloromethyl-2-fluoro-benzene,acmc-209q6a,2-fluoro-4-bromo-chloromethylbenzene
2773361
4-bromo-1-(chloromethyl)-2-fluorobenzene
C1=CC(=C(C=C1Br)F)CCl
Refractive Index 1.5610 to 1.5630 (20°C, 589nm)
Quantity 25g
Color Colorless
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula ClCH2C6H3(F)Br
Formula Weight 223.47
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-fluorobenzyl chloride
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger