Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-bromo-2-fluorobenzylamine, 98%, ACROS Organics™
Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
Specifications
Refractive Index | 1.5630 to 1.5650 (20°C, 589nm) |
Packaging | Glass bottle |
Quantity | 25g |
Color | Colorless to Yellow |
Infrared Spectrum | Authentic |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97.5% min. (GC) |
Linear Formula | NH2CH2C6H3(F)Br |
Formula Weight | 204.04 |
Show More |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously
GHS Signal Word: Danger