Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-fluorobiphenyl, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
41604-19-7
C12H8BrF
251.1
MFCD00051716
HTRNHWBOBYFTQF-UHFFFAOYSA-N
4-bromo-2-fluorobiphenyl,4-bromo-2-fluoro-1,1'-biphenyl,2-fluoro-4-bromo biphenyl,1,1'-biphenyl, 4-bromo-2-fluoro,4-bromo-2-fluorodiphenyl,4-bromo-2-fluoro-1,1-biphenyl,4-bromo-2-fluoro biphenyl,pubchem2652,acmc-209jkv,2-fluoro-4-bromobiphenyl
521063
4-bromo-2-fluoro-1-phenylbenzene
C1=CC=C(C=C1)C2=C(C=C(C=C2)Br)F
41604-19-7
C12H8BrF
251.1
MFCD00051716
HTRNHWBOBYFTQF-UHFFFAOYSA-N
4-bromo-2-fluorobiphenyl,4-bromo-2-fluoro-1,1'-biphenyl,2-fluoro-4-bromo biphenyl,1,1'-biphenyl, 4-bromo-2-fluoro,4-bromo-2-fluorodiphenyl,4-bromo-2-fluoro-1,1-biphenyl,4-bromo-2-fluoro biphenyl,pubchem2652,acmc-209jkv,2-fluoro-4-bromobiphenyl
521063
4-bromo-2-fluoro-1-phenylbenzene
C1=CC=C(C=C1)C2=C(C=C(C=C2)Br)F
Color White to Yellow
Flash Point 113°C
Melting Point 38°C to 42°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 50g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula C6H5C6H3(F)Br
Solubility Information Solubility in water: insoluble
Formula Weight 251.1
Physical Form Crystalline Powder or Chunks
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-fluorobiphenyl
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 255-453-9