Learn More
Accela Chembio IncSupplier Diversity Partner 4-BROMO-2-HYDROXYBENZOIC 25G  DFS Item
Manufacturer:  Accela Chembio IncSupplier Diversity Partner SY00439625G
4-bromo-2-hydroxybenzoic Acid, 25g, 1666-28-0, MFCD06797970, 97+%, Retest Period: 900days, Light Sensitive/moisture Sensitive/nitrogen Or Argon
Catalog No. 50-648-179