Alfa Aesar™ 4-Bromo-2-methoxybenzaldehyde, 97%

Manufacturer: Alfa Aesar™ H55545MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH55545MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 43192-33-2
    Molecular Formula C8H7BrO2
    Formula Weight 215.05
    MDL Number MFCD08276823
    Chemical Name or Material 4-Bromo-2-methoxybenzaldehyde

    Specifications

    Formula Weight 215.05
    Molecular Formula C8H7BrO2
    MDL Number MFCD08276823
    CAS 43192-33-2
    Melting Point 67° to 71°C
    Quantity 250mg
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 4-Bromo-2-methoxybenzaldehyde
    Assay 97%

    Description

    Applications
    4-Bromo-2-methoxybenzaldehyde is used in the building block, synthesis of a nonacid degradable linker for solid-phase synthesis.

    Solubility
    Soluble in water.

    Notes
    Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit